missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
n-Nonanoic acid, 97%
Handla allt Thermo Scientific Chemicals Produkter
Klicka för att se tillgängliga alternativ
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Förpackningsstorlek:
100g
1kg
2.5kg
500g
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Melting Point | 12.5°C |
| Color | Colorless |
| Density | 0.9000g/mL |
| Boiling Point | 254.0°C |
| Flash Point | 114°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Visa mer |
Säkerhet och hantering
Produktbeteckning
- n-Nonanoic acid
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering