Learn More
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride, Thermo Scientific Chemicals
£129.65 - £1388.91
Chemical Identifiers
CAS | 25561-30-2, 75-77-4 |
---|---|
Molecular Formula | C8H18F3NOSi2 |
Molecular Weight (g/mol) | 257.4 |
MDL Number | MFCD00008269 |
InChI Key | XCOBLONWWXQEBS-GHXNOFRVSA-N |
Synonym | bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate |
PubChem CID | 9601896 |
IUPAC Name | trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate |
SMILES | C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10005363
|
Thermo Scientific Acros
389320250 |
25 g |
£129.65
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10330204
|
Thermo Scientific
389321000 |
100 g |
£355.77
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11955861
|
Thermo Scientific Acros
389325000 |
500 g |
£1388.91
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentChemical Identifiers
25561-30-2, 75-77-4 | |
257.4 | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
9601896 | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C |
C8H18F3NOSi2 | |
MFCD00008269 | |
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate |
Specifications
25561-30-2, 75-77-4 | |
0.9740g/mL | |
34°C | |
C8H18F3NOSi2 | |
MFCD00008269 | |
bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate | |
XCOBLONWWXQEBS-GHXNOFRVSA-N | |
trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate | |
9601896 | |
≥98.0% (excluding TMCS), 1.5% max. TMCS | |
N, O-Bis(trimethylsilyl)trifluoroacetamide |
Colorless to Yellow | |
145.0°C to 147.0°C | |
Glass bottle | |
CF3C[=NSi(CH3)3]OSi(CH3)3 | |
0.974 | |
Solubility in water: hydrolysis | |
C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C | |
257.4 | |
257.4 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 247-103-9
- N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.