missing translation for 'onlineSavingsMsg'
Learn More
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%, Thermo Scientific Chemicals Product Code.: 10409100

N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%, Thermo Scientific Chemicals

Product Code. 10409100
100 g, Glass Bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10409100

Brand: Thermo Scientific Acros 168001000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25561-30-2
Molecular Formula C8H18F3NOSi2
Molecular Weight (g/mol) 257.39
MDL Number MFCD00008269
InChI Key XCOBLONWWXQEBS-GHXNOFRVSA-N
Synonym bstfa, n,o-bis trimethylsilyl trifluoroacetamide, acetamide, 2,2,2-trifluoro-o,n-bis trimethylsilyl, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl acetimidate, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanimidate, trimethylsilyl 1z-2,2,2-trifluoro-n-z-trimethylsilyl ethanimidoate #, n,o-bis trimethylsilyl trifluoroacetamide bstfa, trimethylsilyl 2,2,2-trifluoro-n-trimethylsilyl ethanecarboximidate
PubChem CID 9601896
IUPAC Name trimethylsilyl (1Z)-2,2,2-trifluoro-N-trimethylsilylethanimidate
SMILES C[Si](C)(C)N=C(C(F)(F)F)O[Si](C)(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -10.0°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 45.0°C to 50.0°C (14.0 mmHg)
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass Bottle
Linear Formula CF3C[=NSi(CH3)3]OSi(CH3)3
Refractive Index 1.3829 to 1.3849
Quantity 100 g
UN Number 2920
Specific Gravity 0.96
Solubility Information Solubility in water: hydrolyses
Formula Weight 257.39
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material N, O-Bis(trimethylsilyl)trifluoroacetamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466058 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog Number168005000,168000050,168000250,168001000
Lot NumberA0466001 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog Number168005000,168000050,168000250,168001000
Lot NumberA0458230 Certificate TypeCertificate of Analysis Date29/11/2023 Catalog Number168005000,168000050,168000250,168001000
Lot NumberA0446816 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog Number168005000,168000050,168000250,168001000
Lot NumberA0454430 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number168005000,168000050,168000250,168001000
compliance-icons compliance-icons
Product Identifier
  • N,O-Bis(trimethylsilyl)trifluoroacetamide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,O-Bis(trimethylsilyl)trifluoroacetamide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.