missing translation for 'onlineSavingsMsg'
Learn More
n-Octadecylphosphonic acid, 97%, Thermo Scientific Chemicals Product Code.: 11387957

n-Octadecylphosphonic acid, 97%, Thermo Scientific Chemicals

Product Code. 11387957
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11387957

Brand: Thermo Scientific Alfa Aesar 020645.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

n-Octadecylphosphonic acid is used in thermal paper for receipts, adding machines and tickets. It acts as surfactant, emulsifier, dispersants and chelating agents. Further, it is a long chain phosphonic acid compound utilized for proteomics research. In addition to this, it is used to prepare modified mesoporous magnetic nano particles through Lewis acid/base interaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4724-47-4
Molecular Formula C18H39O3P
Molecular Weight (g/mol) 334.481
MDL Number MFCD00015838
InChI Key FTMKAMVLFVRZQX-UHFFFAOYSA-N
Synonym n-octadecylphosphonic acid, phosphonic acid, octadecyl, phosphonic acid, p-octadecyl, n-octadecylphosphonicacid, octadecanephosphonic acid, octadecyl phosphonic acid, octadecyl-phosphonic acid, acmc-209k8h, ksc492m8n, octadecylphosphonic acidShow MoreShow Less
PubChem CID 78451
IUPAC Name octadecylphosphonic acid
SMILES CCCCCCCCCCCCCCCCCCP(=O)(O)O

Specifications

Melting Point 92°C to 98°C
Color White
Linear Formula CH3(CH2)17P(O)(OH)2
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 334.48
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material n-Octadecylphosphonic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC18N16 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number020645.03,020645.06
Lot NumberA29P16 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number020645.03,020645.06
Lot NumberC12W038 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number020645.03,020645.06
Lot NumberF26N12 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number020645.03,020645.06
Lot NumberA29U037 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number020645.03,020645.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Octadecylphosphonic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.