missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11418536
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl 4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol, phenoxy ethanol and propylene glycol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1219-38-1
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00016482
InChI Key RIKCMEDSBFQFAL-UHFFFAOYSA-N
Synonym octylparaben, n-octyl 4-hydroxybenzoate, octyl paraben, octyl p-hydroxybenzoate, p-hydroxybenzoic acid octyl ester, benzoic acid, 4-hydroxy-, octyl ester, p-oxybenzoesaureoctylester, 4-hydroxybenzoic acid n-octyl ester, unii-4uvk431k6b, p-oxybenzoesaureoctylester german
PubChem CID 14642
IUPAC Name octyl 4-hydroxybenzoate
SMILES CCCCCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 50°C to 53°C
Quantity 500 g
Solubility Information Soluble in alcohol,phenoxy ethanol and propylene glycol. Insoluble in water.
Formula Weight 250.34
Percent Purity 98%
Chemical Name or Material n-Octyl 4-hydroxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'