missing translation for 'onlineSavingsMsg'
Learn More

n-Octyl-beta-D-thioglucopyranoside, 98+%, Thermo Scientific Chemicals

Product Code. 15473689
Click to view available options
Quantity:
2.5 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-ionic detergent used for the solubilization of membrane proteins

n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.

Solubility
Soluble in ethanol, water or methanol

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85618-21-9
Molecular Formula C14H28O5S
Molecular Weight (g/mol) 308.433
MDL Number MFCD00012189
InChI Key CGVLVOOFCGWBCS-DGTMBMJNSA-N
Synonym octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
PubChem CID 53720324
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
SMILES CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 128°C
Color White
Quantity 5 g
Beilstein 4182783
Solubility Information Soluble in ethanol, water or methanol
Formula Weight 308.44
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material n-Octyl-beta-D-thioglucopyranoside

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'