Learn More
n-Octyl-beta-D-thioglucopyranoside, 98+%, Thermo Scientific Chemicals
Non-ionic detergent used for the solubilization of membrane proteins
£186.00 - £531.00
Chemical Identifiers
CAS | 85618-21-9 |
---|---|
Molecular Formula | C14H28O5S |
Molecular Weight (g/mol) | 308.433 |
MDL Number | MFCD00012189 |
InChI Key | CGVLVOOFCGWBCS-DGTMBMJNSA-N |
Synonym | octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside |
PubChem CID | 53720324 |
IUPAC Name | (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol |
SMILES | CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15493689
|
Thermo Scientific Alfa Aesar
J61028.88 |
2.5 g |
£186.00
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15473689
|
Thermo Scientific Alfa Aesar
J61028.06 |
5 g |
£221.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15483689
|
Thermo Scientific Alfa Aesar
J61028.09 |
10 g |
£531.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Octyl-beta-D-thioglucopyranoside is used for cell lysis or to solubilise membrane proteins without denaturing them. This is particularly of use in order to crystallise them or to reconstitute them into lipid bilayers. It has been used in a study to assess the crystallization and preliminary x-ray analysis of human plasma apolipoprotein C-I. It has also been used in a study that investigated the identification of Chlamydia pneumoniae proteins.
Solubility
Soluble in ethanol, water or methanol
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
85618-21-9 | |
308.433 | |
CGVLVOOFCGWBCS-DGTMBMJNSA-N | |
53720324 | |
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O |
C14H28O5S | |
MFCD00012189 | |
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol |
Specifications
85618-21-9 | |
White | |
MFCD00012189 | |
octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside | |
CGVLVOOFCGWBCS-DGTMBMJNSA-N | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol | |
53720324 | |
≥98% | |
n-Octyl-beta-D-thioglucopyranoside |
128°C | |
C14H28O5S | |
4182783 | |
Soluble in ethanol, water or methanol | |
CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O | |
308.433 | |
308.44 | |
Crystalline Powder |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only