missing translation for 'onlineSavingsMsg'
Learn More

Melford n-Octylglucoside

Non-ionic detergent for the solubilization and isolation of membrane bound proteins.

£104.00 - £2455.00

Specifications

CAS 29836-26-8
Molecular Formula C14H28O6
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30018552
View Documents
Melford
N02007-50.0
50 g
£2455.00
50g
Please to purchase this item. Need a web account? Register with us today!
30018550
View Documents
Melford
N02007-25.0
25 g
£1235.00
25g
Please to purchase this item. Need a web account? Register with us today!
30018549
View Documents
Melford
N02007-10.0
10 g
£616.00
10g
Please to purchase this item. Need a web account? Register with us today!
30018551
View Documents
Melford
N02007-5.0
5 g
£334.00
5g
Please to purchase this item. Need a web account? Register with us today!
30018548
View Documents
Melford
N02007-1.0
1 g
£104.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Non-ionic detergent for the solubilization and isolation of membrane bound proteins.

Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Specifications

Specifications

29836-26-8
C14H28O6
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.