Learn More
n-Octyltrichlorosilane, 97%, Thermo Scientific Chemicals
£23.12 - £183.92
Chemical Identifiers
CAS | 5283-66-9 |
---|---|
Molecular Formula | C8H17Cl3Si |
Molecular Weight (g/mol) | 247.659 |
MDL Number | MFCD00000488 |
InChI Key | RCHUVCPBWWSUMC-UHFFFAOYSA-N |
Synonym | trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive |
PubChem CID | 21354 |
IUPAC Name | trichloro(octyl)silane |
SMILES | CCCCCCCC[Si](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460188
|
Thermo Scientific Alfa Aesar
B23753.14 |
25 g |
£23.12
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470188
|
Thermo Scientific Alfa Aesar
B23753.22 |
100 g |
£52.76
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480188
|
Thermo Scientific Alfa Aesar
B23753.36 |
500 g |
£183.92
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5283-66-9 | |
247.659 | |
RCHUVCPBWWSUMC-UHFFFAOYSA-N | |
21354 | |
CCCCCCCC[Si](Cl)(Cl)Cl |
C8H17Cl3Si | |
MFCD00000488 | |
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive | |
trichloro(octyl)silane |
Specifications
225°C to 227°C | |
C8H17Cl3Si | |
UN1801 | |
96°C (204°F) | |
RCHUVCPBWWSUMC-UHFFFAOYSA-N | |
trichloro(octyl)silane | |
21354 | |
97% | |
1.447 | |
1.073 |
5283-66-9 | |
MFCD00000488 | |
1744230 | |
trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive | |
CCCCCCCC[Si](Cl)(Cl)Cl | |
247.659 | |
247.67 | |
Irritating | |
Moisture sensitive | |
n-Octyltrichlorosilane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTYLTRICHLOROSILANE
missing translation for 'einecsNumber' : 226-112-1
missing translation for 'rtecsNumber' : VV4700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only