missing translation for 'onlineSavingsMsg'
Learn More
n-Pentane, 95%, technical grade, Thermo Scientific Chemicals Product Code.: 11994401

n-Pentane, 95%, technical grade, Thermo Scientific Chemicals

Product Code. 11994401
5 L, Metal Drum
Click to view available options
Quantity:
5 L
25 L
Packaging:
Metal Drum
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11994401

Brand: Thermo Scientific Acros 434030050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130.0°C
CAS Min % 94.0
Color Colorless
CAS Max % 100.0
Density 0.6260g/mL
Boiling Point 36.0°C
Quantity 5 L
Flash Point −49°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range 95%
Specific Gravity 0.626
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa.s (20°C)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Formula Weight 72.15
Percent Purity 95%
Chemical Name or Material n-Pentane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463998 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number434030250,434030050
Lot NumberA0451995 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number434030250,434030050
Lot NumberA0389943 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number434030250,434030050
Lot NumberA0435422 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog Number434030250,434030050
Lot NumberA0442424 Certificate TypeCertificate of Analysis Date23/05/2022 Catalog Number
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Pentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Pentane, 95%, technical grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.