Learn More
n-Pentane, 99+%, Thermo Scientific Chemicals
£29.64 - £89.37
Chemical Identifiers
CAS | 109-66-0 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009498 |
InChI Key | OFBQJSOFQDEBGM-UHFFFAOYSA-N |
Synonym | n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish |
PubChem CID | 8003 |
ChEBI | CHEBI:37830 |
IUPAC Name | pentane |
SMILES | CCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11984411
|
Thermo Scientific Acros
442991000 |
100 mL |
£29.64
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11974411
|
Thermo Scientific Acros
442990010 |
1 L |
£89.37
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-66-0 | |
72.15 | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
8003 | |
pentane |
C5H12 | |
MFCD00009498 | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polishShow More | |
CHEBI:37830 | |
CCCCC |
Specifications
-130.0°C | |
0.6260g/mL | |
−49°C | |
0.626 | |
CH3(CH2)3CH3 | |
0.25 mPa.s (20°C) | |
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish | |
OFBQJSOFQDEBGM-UHFFFAOYSA-N | |
pentane | |
8003 | |
72.15 | |
Extra Dry over Molecular Sieve |
109-66-0 | |
36.0°C | |
1.358 | |
C5H12 | |
MFCD00009498 | |
0.005% max. | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents | |
CCCCC | |
72.15 | |
CHEBI:37830 | |
≥99% | |
n-Pentane |
Safety and Handling
GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
Danger
- n-Pentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.