missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane-d{12}, 98%(Isotopic)

Product Code. 11384428
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11384428

Brand: Thermo Scientific Alfa Aesar 042340.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2031-90-5 | C5H12 | 84.224 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Density 0.731 g/mL
Flash Point −49°C (−56°F)
Refractive Index 1.358
Sensitivity Light
Quantity 1 g
CAS 2031-90-5
Melting Point -130°C
Boiling Point 36°C
Molecular Formula C5H12
Linear Formula CD3(CD2)3CD3
MDL Number MFCD00190495
UN Number UN1265
Synonym pentane-d12, n-pentane-d12, 2 h?? pentane, pentane-d12, 98 atom % d
InChI Key OFBQJSOFQDEBGM-HYVJACIRSA-N
SMILES CCCCC
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,5-dodecadeuteriopentane
Molecular Weight (g/mol) 84.224
PubChem CID 137286
Formula Weight 84.22
Percent Purity 98%
Chemical Name or Material n-Pentane-d{12}, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.