missing translation for 'onlineSavingsMsg'
Learn More

n-Pentylbenzene, 96%, Thermo Scientific Chemicals

Product Code. 11408770
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-68-1
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00009502
InChI Key PWATWSYOIIXYMA-UHFFFAOYSA-N
Synonym 1-phenylpentane, n-pentylbenzene, amylbenzene, n-amylbenzene, benzene, pentyl, 1-pentylbenzene, 1-phenyl-n-pentane, pentane, 1-phenyl, pentyl-benzene, pentane, phenyl
PubChem CID 10864
IUPAC Name pentylbenzene
SMILES CCCCCC1=CC=CC=C1

Specifications

Melting Point -75°C
Density 0.863
Boiling Point 204°C to 205°C
Flash Point 65°C (149°F)
Refractive Index 1.488
Quantity 5 g
UN Number UN3082
Beilstein 1904359
Merck Index 14,600
Formula Weight 148.25
Percent Purity 96%
Chemical Name or Material n-Pentylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'