Learn More
Thermo Scientific™ N-PER™ Neuronal Protein Extraction Reagent
Collect proteins from all cellular compartments of neuronal tissue and primary cultured neurons with this proprietary, neuronal-specific cell lysis reagent.
Brand: Thermo Scientific™ 87792
Code : PN
Additional Details : Weight : 1.36080kg
Description
Thermo Scientific™ N-PER Neuronal Protein Extraction Reagent is a proprietary cell lysis reagent optimized for efficient extraction of proteins from all cellular compartments of neuronal tissue and primary cultured neurons.
Neuron cell lysates prepared with the N-PER Reagent may be used for enzymatic activity assays (e.g., Phosphatase, Kinase, ATPase Assays), immunoassays (e.g., Western blots, ELISAs, RIAs) and protein purification. The N-PER Lysis Reagent enables protein extraction to be completed in less than 30 minutes and requires mechanical disruption of tissue (e.g., Dounce Homogenization, Polytron). Typical neuronal protein yields are 70-90μg of protein per mg of brain tissue or 300μg of total protein from 106 primary neuronal cells.Highlights:
Optimized – efficient extraction of total neuronal protein, including membrane proteins, from tissue or primary cultured cells
Gentle – preserves protein function without compromising yield
Versatile – can be supplemented with protease inhibitors, reducing or chelating agents or required cofactors
Compatible – extracts are suitable for use with total protein, enzymatic and immunological assays and protein purification methods
Recommended for:
Isolate functional proteins from neuronal tissues or primary neuronal cells; Study enzymatic activity assays specific to neuronal tissue; Generate lysates from neuronal tissues for biomarker analysis
Specifications
Upon receipt store product at 4°C. | |
100 mL | |
Neuronal Protein Extraction Reagent |
NE-PER™ | |
Liquid | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.