missing translation for 'onlineSavingsMsg'
Learn More

N-Phenacylpyridinium bromide, 97%, Thermo Scientific™

£20.30 - £62.20

Chemical Identifiers

CAS 16883-69-5
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.15
InChI Key PXSUMUYPXZEXDT-UHFFFAOYSA-M
Synonym 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
PubChem CID 2798954
IUPAC Name 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
SMILES C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15770879
View Documents Promotion Details
Thermo Scientific Acros
450120100
10 g
£20.30
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15780879
View Documents Promotion Details
Thermo Scientific Acros
450120500
50 g
£62.20
50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

16883-69-5
278.15
1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
C13H12BrNO
PXSUMUYPXZEXDT-UHFFFAOYSA-M
2798954
C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]

Specifications

16883-69-5
Authentic
C13H12BrNO
Solubility in water: soluble.
C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]
278.15
278.15
N-Phenacylpyridinium bromide
193.0°C to 195.0°C
96% min. (Argentometry)
1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
PXSUMUYPXZEXDT-UHFFFAOYSA-M
1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
2798954
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 240-919-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only