missing translation for 'onlineSavingsMsg'
Learn More
N-Phenylbenzylamine, 99%, Thermo Scientific Chemicals Product Code.: 10457022

N-Phenylbenzylamine, 99%, Thermo Scientific Chemicals

Product Code. 10457022
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10457022

Brand: Thermo Scientific Acros 417390050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-32-2
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00003018
InChI Key GTWJETSWSUWSEJ-UHFFFAOYSA-N
Synonym n-phenylbenzylamine, benzylaniline, benzenemethanamine, n-phenyl, phenylbenzylamine, benzylphenylamine, n-monobenzylaniline, aniline, n-benzyl, benzylamine, n-phenyl, benzenamine, n-phenylmethyl, n-benzyl aniline
PubChem CID 66028
IUPAC Name N-benzylaniline
SMILES C1=CC=C(C=C1)CNC2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 34°C to 38°C
Color Beige-Yellow to Colorless
Density 1.0600g/mL
Boiling Point 306°C to 307°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NHC6H5
Quantity 5 g
Beilstein 12, 1023
Merck Index 15, 1129
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble in water. Other solubilities: very soluble in ethanol,ether,soluble in chloroform
Formula Weight 183.25
Percent Purity 99%
Physical Form Crystalline Powder or Low Melting Mass
Chemical Name or Material N-Phenylbenzylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0409248 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog Number
Lot NumberA0458475 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number417395000,417391000,417390250,417390050
Lot NumberA0404482 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0447707 Certificate TypeCertificate of Analysis Date21/11/2022 Catalog Number
Lot NumberA0447708 Certificate TypeCertificate of Analysis Date21/11/2022 Catalog Number
Product Identifier
  • N-Phenylbenzylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Phenylbenzylamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.