missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyldiethanolamine, 97%, Thermo Scientific Chemicals

Product Code. 11318835
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

Solubility
Soluble in water 3g/L @ 20˚C.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-07-0
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00002845
InChI Key OJPDDQSCZGTACX-UHFFFAOYSA-N
Synonym n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
PubChem CID 8416
IUPAC Name 2-[N-(2-hydroxyethyl)anilino]ethanol
SMILES OCCN(CCO)C1=CC=CC=C1

Specifications

Melting Point 55°C to 59°C
Density 1.1
pH 8.2
Boiling Point 270°C
Flash Point 178°C (352°F)
Odor Amine-like
Linear Formula C6H5N(CH2CH2OH)2
Quantity 2500 g
Beilstein 2096832
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water 3g/L @ 20?C.
Formula Weight 181.23
Percent Purity 97%
Chemical Name or Material N-Phenyldiethanolamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'