Learn More
N-Phenylethanolamine, 98%, Thermo Scientific Chemicals
£27.74 - £93.36
Chemical Identifiers
CAS | 122-98-5 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.182 |
MDL Number | MFCD00002832 |
InChI Key | MWGATWIBSKHFMR-UHFFFAOYSA-N |
Synonym | 2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700 |
PubChem CID | 31235 |
IUPAC Name | 2-anilinoethanol |
SMILES | C1=CC=C(C=C1)NCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309759
|
Thermo Scientific Alfa Aesar
B22131.36 |
500 g |
£27.74
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399749
|
Thermo Scientific Alfa Aesar
B22131.0E |
2500 g |
£93.36
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5'-diphospho-glucuronosyltransferase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Phenylethanolamine is used as an intermediate for dyes and other organic compounds. Further, it is used as a substrate for human olfactory uridine 5′-diphospho-glucuronosyltransferase.
Solubility
Slightly miscible with water.
Notes
Incompatible with acids, acid anhydrides, acid chlorides and chloroformates.
Identifiants chimiques
122-98-5 | |
137.182 | |
MWGATWIBSKHFMR-UHFFFAOYSA-N | |
31235 | |
C1=CC=C(C=C1)NCCO |
C8H11NO | |
MFCD00002832 | |
2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700 | |
2-anilinoethanol |
Spécification
122-98-5 | |
1.087 | |
284°C to 286°C | |
Amine-like | |
1.579 | |
UN2810 | |
2-phenylamino ethanol, n-phenylethanolamine, n-2-hydroxyethyl aniline, ethanol, 2-phenylamino, 2-phenylamino ethan-1-ol, ethanol, 2-anilino, beta-anilinoethanol, aniline, n-2-hydroxyethyl, n-2-hydroxyethyl phenylamine, emery 5700 | |
MWGATWIBSKHFMR-UHFFFAOYSA-N | |
2-anilinoethanol | |
31235 | |
98% |
-30°C | |
10.2 | |
153°C (307°F) | |
C8H11NO | |
MFCD00002832 | |
774672 | |
Slightly miscible with water. | |
C1=CC=C(C=C1)NCCO | |
137.182 | |
137.18 | |
N-Phenylethanolamine |
Sécurité et traitement
GHS H Statement
H310-H351-H373-H319-H317
Fatal in contact with skin.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause an allergic skin reaction.
P264b-P280-P302+P352-P305+P351+P338-P312-P361-P363-P501c
H311-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 204-588-1
missing translation for 'rtecsNumber' : KJ7175000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.