missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Phenylglycine, 93%
£31.90 - £84.60
Chemical Identifiers
CAS | 103-01-5 |
---|---|
Molecular Formula | C8H8NNaO2 |
Molecular Weight (g/mol) | 173.15 |
MDL Number | MFCD00014009 |
InChI Key | YEMGQZDWLLBIEY-UHFFFAOYSA-M |
Synonym | n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine |
PubChem CID | 66025 |
ChEBI | CHEBI:55477 |
IUPAC Name | 2-anilinoacetic acid |
SMILES | [Na+].[O-]C(=O)CNC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10349390
|
Thermo Scientific Chemicals
276080250 |
25 g | Glass bottle |
£31.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10083180
|
Thermo Scientific Chemicals
276081000 |
100 g | Glass bottle |
£84.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103-01-5 | |
173.15 | |
YEMGQZDWLLBIEY-UHFFFAOYSA-M | |
66025 | |
2-anilinoacetic acid |
C8H8NNaO2 | |
MFCD00014009 | |
n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine | |
CHEBI:55477 | |
[Na+].[O-]C(=O)CNC1=CC=CC=C1 |
Specifications
103-01-5 | |
100.0 | |
Brown-Yellow to Ochre | |
91% min. (HPLC) | |
C6H5NHCH2CO2H | |
12, 468 | |
n-phenylglycine, anilinoacetic acid, 2-phenylamino acetic acid, glycine, n-phenyl, glycine, phenyl, n-phenylglycin, n-phenylaminoacetic acid, phenylamino acetic acid, h-phenylgly-oh, n-phenyl glycine | |
YEMGQZDWLLBIEY-UHFFFAOYSA-M | |
2-anilinoacetic acid | |
66025 | |
151.17 | |
Fine Powder |
91.0 | |
121.0°C to 127.0°C | |
Authentic | |
C8H8NNaO2 | |
MFCD00014009 | |
15, 7403 | |
Solubility in water: moderately soluble. Other solubilities: sparingly soluble in ether | |
[Na+].[O-]C(=O)CNC1=CC=CC=C1 | |
173.15 | |
CHEBI:55477 | |
93% | |
N-Phenylglycine, 93% |
Safety and Handling
missing translation for 'einecsNumber' : 203-070-2
missing translation for 'tsca' : TSCA
RUO – Research Use Only