missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylrhodanine, 97+%, Thermo Scientific™

Product Code. 10269093 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10269093

Brand: Thermo Scientific Acros 391390050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1457-46-1
Molecular Formula C9H7NOS2
Molecular Weight (g/mol) 209.29
MDL Number MFCD00100196
InChI Key DVRWEKGUWZINTQ-UHFFFAOYSA-N
Synonym 3-phenylrhodanine, n-phenylrhodanine, 3-phenylrhodanin, rhodanine, 3-phenyl, 4-thiazolidinone, 3-phenyl-2-thioxo, 3-phenylrodanin, n-phenyl-rhodanine, 3-phenyl-2-thioxo-1,3-thiazolidin-4-one, 3-phenyl-2-thioxo-thiazolidin-4-one, 3-phenyl-rhodanin german
PubChem CID 73822
IUPAC Name 3-phenyl-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)N(C(=S)S1)C2=CC=CC=C2

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 190°C to 194°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 209.29
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylrhodanine
Show More Show Less
compliance-icons
Product Identifier
  • N-Phenylrhodanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.