missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propylbenzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.
Solubility
Miscible with alcohol, ether, acetone, benzene, and petroleum ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
n-Propylbenzene is used to prepare benzoic acid. It is employed as a solvent for gas chromatography. It is also used as a solvent and an intermediate in organic synthesis. Further, it is used in fuels and fuel additives.
Solubility
Miscible with alcohol, ether, acetone, benzene, and petroleum ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | °C to 100°C |
Density | 0.862 |
Boiling Point | 159°C to 160°C |
Flash Point | 47°C (116°F) |
Assay Percent Range | 0.98 |
Refractive Index | 1.491 |
Quantity | 100 g |
UN Number | UN1993 |
Beilstein | 1903006 |
Merck Index | 14,7844 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'