missing translation for 'onlineSavingsMsg'
Learn More

N-[(S)-alpha-(4-Chlorophenyl)benzyl]formamide, 98%, Thermo Scientific™

Product Code. 15457948
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474654-19-8
Molecular Formula C14H12ClNO
Molecular Weight (g/mol) 245.71
InChI Key HDGOSEHHSVUCBO-UHFFFAOYNA-N
Synonym n-s-4-chlorophenyl phenyl methyl formamide, n-s-4-chlorophenyl phenylmethyl-formamide

Specifications

Quantity 1 g
Formula Weight 245.7
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-[(S)-alpha-(4-Chlorophenyl)benzyl]formamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'