missing translation for 'onlineSavingsMsg'
Learn More
N-tert-Butoxycarbonyl-1,6-hexanediamine, 95%, Thermo Scientific Chemicals Product Code.: 10560404

N-tert-Butoxycarbonyl-1,6-hexanediamine, 95%, Thermo Scientific Chemicals

Product Code. 10560404
5 g, Plastic Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10560404

Brand: Thermo Scientific Acros 301830050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51857-17-1
Molecular Formula C11H24N2O2
Molecular Weight (g/mol) 216.33
MDL Number MFCD00671489
InChI Key RVZPDKXEHIRFPM-UHFFFAOYSA-N
Synonym n-boc-1,6-diaminohexane, n-boc-1,6-hexanediamine, tert-butyl n-6-aminohexyl carbamate, tert-butyl 6-aminohexyl carbamate, n-tert-butoxycarbonyl-1,6-diaminohexane, n-tert-butoxycarbonyl-1,6-hexanediamine, tert-butyl 6-aminohexylcarbamate, boc-1,6-diaminohexane hydrochloride, 6-tert-butoxycarbonylamino hexylamine, n-6-aminohexyl carbamic acid tert-butyl esterShow MoreShow Less
PubChem CID 2733170
IUPAC Name tert-butyl N-(6-aminohexyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCCCN

Specifications

Boiling Point 106°C to 110°C (0.3mmHg)
CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic Bottle
Refractive Index 1.4590 to 1.461
UN Number 2735
Solubility Information Solubility in water: .. Other solubilities: soluble in ether,ch2cl2
Quantity 5 g
Flash Point 125°C
Formula Weight 216.33
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material N-tert-Butoxycarbonyl-1, 6-hexanediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0407667 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog Number301830050,301830010
Lot NumberA0403174 Certificate TypeCertificate of Analysis Date31/05/2021 Catalog Number301830050,301830010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-tert-Butoxycarbonyl-1,6-hexanediamine, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.