missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-tert-Butoxycarbonyl-L-prolinol, 98%, Thermo Scientific Chemicals
£71.30 - £244.00
Chemical Identifiers
CAS | 69610-40-8 |
---|---|
Molecular Formula | C10H19NO3 |
Molecular Weight (g/mol) | 201.26 |
MDL Number | MFCD00066232 |
InChI Key | BFFLLBPMZCIGRM-QMMMGPOBSA-N |
Synonym | n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol |
PubChem CID | 643448 |
IUPAC Name | tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCCC1CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10357811
|
Thermo Scientific
344560050 |
5 g | Glass bottle |
£71.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135313
|
Thermo Scientific Acros
344560250 |
25 g | Glass bottle |
£244.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
69610-40-8 | |
201.26 | |
BFFLLBPMZCIGRM-QMMMGPOBSA-N | |
643448 | |
CC(C)(C)OC(=O)N1CCCC1CO |
C10H19NO3 | |
MFCD00066232 | |
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol | |
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
Specifications
69610-40-8 | |
100.0 | |
White | |
97.5% min. (GC) | |
MFCD00066232 | |
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol | |
BFFLLBPMZCIGRM-QMMMGPOBSA-N | |
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate | |
643448 | |
98% | |
N-tert-Butoxycarbonyl-L-prolinol |
97.5 | |
60°C to 64°C | |
Authentic | |
C10H19NO3 | |
-52 | |
Solubility in water: insoluble in water | |
CC(C)(C)OC(=O)N1CCCC1CO | |
201.26 | |
201.26 | |
Crystalline Powder |
RUO – Research Use Only