missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-tert-Butylacetamide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | 95°C to 99°C |
Quantity | 10 g |
Beilstein | 1699507 |
Solubility Information | Soluble in water. |
Formula Weight | 115.18 |
Percent Purity | 98% |
Chemical Name or Material | N-tert-Butylacetamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'