Learn More
N-tert-Butylcyclohexylamine, 97%, Thermo Scientific Chemicals
£38.34 - £163.12
Chemical Identifiers
CAS | 51609-06-4 |
---|---|
Molecular Formula | C10H21N |
Molecular Weight (g/mol) | 155.29 |
InChI Key | MTEWAFVECQBILW-UHFFFAOYSA-N |
Synonym | n-tert-butylcyclohexylamine, n-tert-butyl cyclohexanamine, n-t-butylcyclohexylamine, tert-butyl-cyclohexylamine, tert-butylcyclohexyl amine, tert-butyl cyclohexylamine, tert.-butyl-cyclohexyl-amine, n-tertbutyl-n-cyclohexyl amine, n-tert-butyl-n-cyclohexyl amine, n-1,1-dimethylethyl cyclohexanamine |
PubChem CID | 103920 |
IUPAC Name | N-tert-butylcyclohexanamine |
SMILES | CC(C)(C)NC1CCCCC1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51609-06-4 | |
155.29 | |
n-tert-butylcyclohexylamine, n-tert-butyl cyclohexanamine, n-t-butylcyclohexylamine, tert-butyl-cyclohexylamine, tert-butylcyclohexyl amine, tert-butyl cyclohexylamine, tert.-butyl-cyclohexyl-amine, n-tertbutyl-n-cyclohexyl amine, n-tert-butyl-n-cyclohexyl amine, n-1,1-dimethylethyl cyclohexanamine | |
N-tert-butylcyclohexanamine |
C10H21N | |
MTEWAFVECQBILW-UHFFFAOYSA-N | |
103920 | |
CC(C)(C)NC1CCCCC1 |
Specifications
51609-06-4 | |
100.0 | |
0.8340g/mL | |
50°C | |
96% min. (GC) | |
0.834 | |
MTEWAFVECQBILW-UHFFFAOYSA-N | |
N-tert-butylcyclohexanamine | |
103920 | |
97% | |
N-tert-Butylcyclohexylamine |
96.0 | |
Colorless to Yellow | |
172°C to 174°C | |
Authentic | |
C10H21N | |
n-tert-butylcyclohexylamine, n-tert-butyl cyclohexanamine, n-t-butylcyclohexylamine, tert-butyl-cyclohexylamine, tert-butylcyclohexyl amine, tert-butyl cyclohexylamine, tert.-butyl-cyclohexyl-amine, n-tertbutyl-n-cyclohexyl amine, n-tert-butyl-n-cyclohexyl amine, n-1,1-dimethylethyl cyclohexanamine | |
CC(C)(C)NC1CCCCC1 | |
155.29 | |
155.29 | |
Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapour.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 257-320-
- N-tert-Butylcyclohexylamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only