missing translation for 'onlineSavingsMsg'
Learn More

n-Tetracosane, 99%

Product Code. 11328978
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11328978

Brand: Thermo Scientific Alfa Aesar 042462.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 646-31-1 | C24H50 | 338.664 g/mol

n-Tetracosane is a suitable solvent used in the synthesis of ZnS nanoparticle. It is also employed as the wax component to study the reduction in pour point of hydrocarbon solvents containing wax crystals on addition of polymer additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-31-1
Molecular Formula C24H50
Molecular Weight (g/mol) 338.664
MDL Number MFCD00009352
InChI Key POOSGDOYLQNASK-UHFFFAOYSA-N
Synonym n-tetracosane, lignocerane, unii-yq5h1m1d7i, tricosane, methyl, yq5h1m1d7i, tetracosane, analytical standard, tetracosane, acmc-1b0gq, n-tetracosane 5g, ksc357o9b
PubChem CID 12592
ChEBI CHEBI:32936
IUPAC Name tetracosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 49°C to 52°C
Density 0.799
Boiling Point 391°C
Flash Point >110°C (230°F)
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 338.66
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material n-Tetracosane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.