missing translation for 'onlineSavingsMsg'
Learn More
N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals Product Code.: 11986711

N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 11986711
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11986711

Brand: Thermo Scientific Acros 441020050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 365564-11-0
Molecular Formula C19H36BNO2Si
Molecular Weight (g/mol) 349.40
MDL Number MFCD08063117
InChI Key GWFIZBYDIHGZRJ-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris 1-methylethyl silyl-1h-pyrrole, 3-tetramethyl-1,3,2-dioxaborolan-2-yl-1-tris propan-2-yl silyl-1h-pyrrole, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl-1h-pyrrole, 3-pinacolateboryl-1-tips-pyrrole, 1-triisopropylsilyl-1h-pyrrol-3-ylboronic acid pinacol ester, 1-triisopropylsilyl-1h-pyrrole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1-triisopropylsilyl pyrrole, pubchem18460, pyrrole-3-boronic acid, pinacol ester;, n-tips pyrrole-3-boronic acid pinacol esterShow MoreShow Less
PubChem CID 11450924
IUPAC Name tri(propan-2-yl)-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrrol-1-yl]silane
SMILES CC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 70°C to 72°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 349.4
Percent Purity 95%
Physical Form Powder or Solid
Chemical Name or Material N-TIPS pyrrole-3-boronic acid pinacol ester
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number337192 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number441020010,441020050
compliance-icons
Product Identifier
  • N-TIPS Pyrrole-3-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.