missing translation for 'onlineSavingsMsg'
Learn More

n-Tricosane, 99%, Thermo Scientific Chemicals

£45.83 - £529.64

Chemical Identifiers

CAS 638-67-5
Molecular Formula C23H48
Molecular Weight (g/mol) 324.637
MDL Number MFCD00009350
InChI Key FIGVVZUWCLSUEI-UHFFFAOYSA-N
Synonym n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
PubChem CID 12534
ChEBI CHEBI:32934
IUPAC Name tricosane
SMILES CCCCCCCCCCCCCCCCCCCCCCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11431978
View Documents
Thermo Scientific Alfa Aesar
A14577.06
5 g
£45.83
5g
Please to purchase this item. Need a web account? Register with us today!
11441978
View Documents
Thermo Scientific Alfa Aesar
A14577.14
25 g
£171.86
25g
Please to purchase this item. Need a web account? Register with us today!
11451978
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14577.22
100 g
£529.64
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

n-Tricosane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tricosane is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

638-67-5
324.637
FIGVVZUWCLSUEI-UHFFFAOYSA-N
12534
tricosane
C23H48
MFCD00009350
n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
CHEBI:32934
CCCCCCCCCCCCCCCCCCCCCCC

Specifications

638-67-5
0.7969
>110°C (230°F)
1.4468
1756000
Insoluble in water.
CCCCCCCCCCCCCCCCCCCCCCC
324.637
CHEBI:32934
99%
47°C to 50°C
199°C to 200°C (3 mmHg)
C23H48
MFCD00009350
n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
FIGVVZUWCLSUEI-UHFFFAOYSA-N
tricosane
12534
324.64
n-Tricosane
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 211-347-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only