missing translation for 'onlineSavingsMsg'
Learn More

N-(alpha,alpha,alpha-Trifluoro-p-tolyl)piperazine, 98%

Product Code. p-2691830 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10018941

Brand: Thermo Scientific Acros 367120010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 30459-17-7 | C11H13F3N3 | 230.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30459-17-7
Molecular Formula C11H13F3N3
Molecular Weight (g/mol) 230.23
MDL Number MFCD00040765
InChI Key IBQMAPSJLHRQPE-UHFFFAOYSA-N
Synonym 1-4-trifluoromethylphenyl piperazine, 1-4-trifluoromethyl phenyl piperazine, piperazine, 1-4-trifluoromethyl phenyl, 1-4-trifluoromethylphenyl-piperazine, 1-4-trifluoromethyl-phenyl-piperazine, n-4-trifluoromethylphenyl piperazine, 4-trifluoromethyl phenyl piperazine, zlchem 639, acmc-209hfy
PubChem CID 121718
IUPAC Name 1-[4-(trifluoromethyl)phenyl]piperazine
SMILES C1CN(CCN1)C2=CC=C(C=C2)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89.0°C to 92.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 230.23
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material N-(α, α, α-Trifluoro-p-tolyl)piperazine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-(a,a,a-Trifluoro-p-tolyl)piperazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.