missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)acetamide, 97%, Thermo Scientific Chemicals

Product Code. 10286880
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13435-12-6
Molecular Formula C5H13NOSi
Molecular Weight (g/mol) 131.25
MDL Number MFCD00008671
InChI Key LWFWUJCJKPUZLV-UHFFFAOYSA-N
Synonym n-trimethylsilyl acetamide, trimethylsilylacetamide, acetamide, n-trimethylsilyl, tms acetamide, acetylamino trimethylsilane, trimethylsilyl acetamide, acetamide, tms derivative, n-trimethylsilyl-acetamide, n-tms-acetamide, n-trimethylsilylacetoamide
PubChem CID 25989
IUPAC Name N-trimethylsilylacetamide
SMILES CC(=O)N[Si](C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 46°C
Color White to Yellow
Boiling Point 84°C (18.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3CONHSi(CH3)3
Quantity 25 g
Fieser 01,1235; 16,178
Solubility Information Solubility in water: reacts
Formula Weight 131.25
Percent Purity 97%
Physical Form Crystalline Mass
Chemical Name or Material N-(Trimethylsilyl)acetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-(Trimethylsilyl)acetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'