missing translation for 'onlineSavingsMsg'
Learn More

N-(Trimethylsilyl)diethylamine, 97%, Thermo Scientific Chemicals

Product Code. 11403667
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 996-50-9
Molecular Formula C7H19NSi
Molecular Weight (g/mol) 145.32
MDL Number MFCD00009040
InChI Key JOOMLFKONHCLCJ-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
PubChem CID 70454
ChEBI CHEBI:85070
IUPAC Name N-ethyl-N-trimethylsilylethanamine
SMILES CCN(CC)[Si](C)(C)C

Specifications

Boiling Point 125°C to 126°C
Melting Point -10°C
Quantity 100 g
UN Number UN2733
Beilstein 635718
Flash Point 10°C (50°F)
Solubility Information Hydrolyzes in water.
Formula Weight 145.32
Percent Purity 98%
Odor Amine-like
Refractive Index 1.411
Sensitivity Moisture sensitive
Density 0.76
Chemical Name or Material N-(Trimethylsilyl)diethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'