Learn More
N-(Trimethylsilyl)diethylamine, 97%, Thermo Scientific Chemicals
£30.25
Chemical Identifiers
CAS | 996-50-9 |
---|---|
Molecular Formula | C7H19NSi |
Molecular Weight (g/mol) | 145.32 |
MDL Number | MFCD00009040 |
InChI Key | JOOMLFKONHCLCJ-UHFFFAOYSA-N |
Synonym | n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine |
PubChem CID | 70454 |
ChEBI | CHEBI:85070 |
IUPAC Name | N-ethyl-N-trimethylsilylethanamine |
SMILES | CCN(CC)[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11493657
|
Thermo Scientific Alfa Aesar
A18149.14 |
25 g |
£30.25
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403667
|
Thermo Scientific Alfa Aesar
A18149.22 |
100 g |
N/A
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used to derivative polar organic compounds. N,N-Diethyl(trimethylsilyl)amine is a chemical reagent with wide application in organic chemistry and used in stereo specific addition reactions as well as in SN2 nucleophilic substitutions.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
996-50-9 | |
145.32 | |
JOOMLFKONHCLCJ-UHFFFAOYSA-N | |
70454 | |
N-ethyl-N-trimethylsilylethanamine |
C7H19NSi | |
MFCD00009040 | |
n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine | |
CHEBI:85070 | |
CCN(CC)[Si](C)(C)C |
Specifications
125°C to 126°C | |
996-50-9 | |
MFCD00009040 | |
635718 | |
n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine | |
JOOMLFKONHCLCJ-UHFFFAOYSA-N | |
N-ethyl-N-trimethylsilylethanamine | |
70454 | |
145.32 | |
Amine-like | |
Moisture sensitive | |
N-(Trimethylsilyl)diethylamine |
-10°C | |
C7H19NSi | |
UN2733 | |
10°C (50°F) | |
Hydrolyzes in water. | |
CCN(CC)[Si](C)(C)C | |
145.32 | |
CHEBI:85070 | |
98% | |
1.411 | |
0.76 |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 213-637-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- N-(Trimethylsilyl)diethylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : N-(Trimethylsilyl)diethylamine
RUO – Research Use Only