Learn More
N-(Trimethylsilyl)imidazole, 97%, Thermo Scientific Chemicals
N-(Trimethylsilyl)imidazole, 97%, ACROS Organics, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 93 deg.C to 94 deg.C (13.0mmHg), Colorless to Yellow, Melting Point: -42 deg.C, Molecular Weight: 140.26, Percent Purity: 97%, Assay Percent Range: 96% min. (GC), CAS: 18156-74-6
£46.20 - £303.00
Chemical Identifiers
CAS | 18156-74-6 |
---|---|
Molecular Formula | C6H12N2Si |
Molecular Weight (g/mol) | 140.26 |
MDL Number | MFCD00005280 |
InChI Key | YKFRUJSEPGHZFJ-UHFFFAOYSA-N |
Synonym | n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl |
PubChem CID | 28925 |
ChEBI | CHEBI:85063 |
IUPAC Name | imidazol-1-yl(trimethyl)silane |
SMILES | C[Si](C)(C)N1C=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10317540
|
Thermo Scientific Acros
167130250 |
25 g | Glass bottle |
£46.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10677522
|
Thermo Scientific Acros
167131000 |
100 g | Glass bottle |
£138.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10475555
|
Thermo Scientific Acros
167132500 |
250 g | Glass bottle |
£303.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagentChemical Identifiers
18156-74-6 | |
140.26 | |
YKFRUJSEPGHZFJ-UHFFFAOYSA-N | |
28925 | |
imidazol-1-yl(trimethyl)silane |
C6H12N2Si | |
MFCD00005280 | |
n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl | |
CHEBI:85063 | |
C[Si](C)(C)N1C=CN=C1 |
Specifications
18156-74-6 | |
100.0 | |
Colorless to Yellow | |
93°C to 94°C (13.0 mmHg) | |
Authentic | |
C6H12N2Si | |
MFCD00005280 | |
0.95 | |
Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents | |
C[Si](C)(C)N1C=CN=C1 | |
140.26 | |
CHEBI:85063 | |
140.26 | |
Liquid |
96.0 | |
-42°C | |
0.9500g/mL | |
5°C | |
96% min. (GC) | |
1.4750 to 1.478 | |
07,399; 11,575; 01,337; 02,177 | |
n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl | |
YKFRUJSEPGHZFJ-UHFFFAOYSA-N | |
imidazol-1-yl(trimethyl)silane | |
28925 | |
2.5 mm2/s (25°C) | |
97% | |
N-(Trimethylsilyl)imidazole |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 242-040-3
- N-(Trimethylsilyl)imidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only