Learn More
N-[Tris(hydroxymethyl)methyl]acrylamide, 99%, Thermo Scientific Chemicals
£38.10 - £516.00
Chemical Identifiers
CAS | 13880-05-2 |
---|---|
Molecular Formula | C7H13NO4 |
Molecular Weight (g/mol) | 175.18 |
MDL Number | MFCD00081045 |
InChI Key | MVBJSQCJPSRKSW-UHFFFAOYSA-N |
Synonym | n-tris hydroxymethyl methyl acrylamide, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide, tris-acrylamide, n-acryloyltris hydroxymethyl aminomethane, n-acryloyl-tris hydroxymethyl aminomethane, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide, zlchem 121, acryloylaminomethylidynetrimethanol, n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide |
PubChem CID | 83788 |
IUPAC Name | N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide |
SMILES | OCC(CO)(CO)NC(=O)C=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11469887
|
Thermo Scientific Alfa Aesar
B22854.06 |
5 g |
£38.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479887
|
Thermo Scientific Alfa Aesar
B22854.14 |
25 g |
£142.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489887
|
Thermo Scientific Alfa Aesar
B22854.22 |
100 g |
£516.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.
Solubility
Soluble in water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.
Chemical Identifiers
13880-05-2 | |
175.18 | |
MVBJSQCJPSRKSW-UHFFFAOYSA-N | |
83788 | |
OCC(CO)(CO)NC(=O)C=C |
C7H13NO4 | |
MFCD00081045 | |
n-tris hydroxymethyl methyl acrylamide, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide, tris-acrylamide, n-acryloyltris hydroxymethyl aminomethane, n-acryloyl-tris hydroxymethyl aminomethane, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide, zlchem 121, acryloylaminomethylidynetrimethanol, n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide | |
N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide |
Specifications
13880-05-2 | |
C7H13NO4 | |
MFCD00081045 | |
n-tris hydroxymethyl methyl acrylamide, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide, tris-acrylamide, n-acryloyltris hydroxymethyl aminomethane, n-acryloyl-tris hydroxymethyl aminomethane, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide, zlchem 121, acryloylaminomethylidynetrimethanol, n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide | |
MVBJSQCJPSRKSW-UHFFFAOYSA-N | |
N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide | |
83788 | |
99% |
134°C to 138°C | |
CH2=CHCONHC(CH2OH)3 | |
Hygroscopic; Light sensitive | |
Soluble in water. | |
OCC(CO)(CO)NC(=O)C=C | |
175.18 | |
175.18 | |
N-[Tris(hydroxymethyl)methyl]acrylamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 237-645-4
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only