missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Undecane, 99.7%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Color | Colorless |
Density | 0.744 at 25°C (water = 1) |
Assay Percent Range | 0.997 |
Quantity | 500 mL |
Formula Weight | 156.3 |
Physical Form | Liquid |
Chemical Name or Material | n-Undecane |
Safety and Handling

Product Identifier
- undecane
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : undecane
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
n-Undecane, 99.7%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction