missing translation for 'onlineSavingsMsg'
Learn More

N2-Methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine, 97%, Thermo Scientific™

£17.30 - £120.00

Chemical Identifiers

CAS 25366-22-7
Molecular Formula C4H4F3N3S
Molecular Weight (g/mol) 183.15
MDL Number MFCD00042321
InChI Key ARAFQFMXQDBEPU-UHFFFAOYSA-N
Synonym n-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazole, n2-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazol, 1,3,4-thiadiazol-2-amine, n-methyl-5-trifluoromethyl, 1,3,4-thiadiazol-2-amine,n-methyl-5-trifluoromethyl, methyl 5-trifluoromethyl 1,3,4-thiadiazol-2-yl amine, maybridge1_002895, methyl-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-amine
PubChem CID 2775323
IUPAC Name N-methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
SMILES CNC1=NN=C(S1)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10334242
View Documents
Thermo Scientific Maybridge
CD05460DA
1 g Amber Glass Bottle
£17.30
1g
Please to purchase this item. Need a web account? Register with us today!
10602643
View Documents
Thermo Scientific Maybridge
10 g Amber Glass Bottle
£120.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

25366-22-7
183.15
ARAFQFMXQDBEPU-UHFFFAOYSA-N
2775323
CNC1=NN=C(S1)C(F)(F)F
C4H4F3N3S
MFCD00042321
n-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazole, n2-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazol, 1,3,4-thiadiazol-2-amine, n-methyl-5-trifluoromethyl, 1,3,4-thiadiazol-2-amine,n-methyl-5-trifluoromethyl, methyl 5-trifluoromethyl 1,3,4-thiadiazol-2-yl amine, maybridge1_002895, methyl-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-amine
N-methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine

Specifications

25366-22-7
100.0
C4H4F3N3S
n-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazole, n2-methyl-5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-methylamino-5-trifluoromethyl-1,3,4-thiadiazol, 1,3,4-thiadiazol-2-amine, n-methyl-5-trifluoromethyl, 1,3,4-thiadiazol-2-amine,n-methyl-5-trifluoromethyl, methyl 5-trifluoromethyl 1,3,4-thiadiazol-2-yl amine, maybridge1_002895, methyl-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-amine
CNC1=NN=C(S1)C(F)(F)F
183.15
183.16
N2-Methyl-5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-amine
96.5
97%
MFCD00042321
ARAFQFMXQDBEPU-UHFFFAOYSA-N
N-methyl-5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
2775323
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications