missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N6-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-fluoro-2'-deoxyadenosine-3'-(2-cyanoethyl diisopropylphosphoramidite), 98%
Click to view available options
Quantity:
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
CAS | 136834-22-5 |
Color | White |
Molecular Formula | C47H51FN7O7P |
MDL Number | MFCD12912392 |
Quantity | 1 g |
Synonym | 2r,3r,4r,5r-5-6-benzamido-9h-purin-9-yl-2-bis 4-methoxyphenyl phenyl methoxy methyl-4-fluorotetrahydrofuran-3-yl 2-cyanoethyl diisopropylphosphoramidite, dmt-2'fluoro-da bz phosphoramidite, dmt-2'fluoro-da bz phosphoramidite, configured for abi, dmt-2'fluoro-da bz phosphoramidite, configured for perkinelmer, configured for polygen, 2'-f-bz-ra, dmt-2'fluoro-da bz amidite, 2'-fluoro-n6-bz-5'-o-dmt-2'-da-3'-ce-phosphoramidite, dmt-2'fluoro-da bz phosphoramidite, configured for ??kta r and oligopilot r, n-9-2r,3r,4r,5r-5-bis 4-methoxyphenyl phenyl methoxy methyl-4-2-cyanoethoxy diisopropylamino phosphanyl oxy-3-fluorooxolan-2-yl purin-6-yl benzamide, n-benzoyl-5'-o-bis 4-methoxyphenyl phenylmethyl-2'-deoxy-2'-fluoroadenosine 3'-2-cyanoethyl n,n-bis 1-methylethyl phosphoramidite |
InChI Key | NZDMBUCNULHYQM-JRJFZAHINA-N |
SMILES | COC1=CC=C(C=C1)C(O[C@@H]1[C@@H](COP(OCCC#N)N(C(C)C)C(C)C)O[C@H]([C@@H]1F)N1C=NC2=C(NC(=O)C3=CC=CC=C3)N=CN=C12)(C1=CC=CC=C1)C1=CC=C(OC)C=C1 |
IUPAC Name | N-[9-[(2R,3R,4R,5R)-5-[[bis(4-methoxyphenyl)-phenylmethoxy]methyl]-4-[2-cyanoethoxy-[di(propan-2-yl)amino]phosphanyl]oxy-3-fluorooxolan-2-yl]purin-6-yl]benzamide |
Molecular Weight (g/mol) | 875.94 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'