Learn More
Invitrogen™ NA-Star™ Detection Microplates
Solid white 96-well microplates, supplied with covers, optimized for use with the chemiluminescent assay reagents. Thermo Scientific NA-Star NA-Star Detection Microplates are ideal for use with other chemiluminescent assays that do not require a specific
Brand: Invitrogen™ 4374349
This item is not returnable.
View return policy
Description
Ideal for use with other chemiluminescent assays that do not require a specific surface chemistry or coating. They do provide a high binding surface that can be coated with specific capture reagents for use with immunoassay⁄capture assay applications, if desired.Specifications
No Barcode | |
Contains 10 plates with covers | |
NA-Star NovaBright | |
None | |
Room Temperature |
Standard | |
Colorimetric | |
Luminometer (Microplate) | |
10 plates |
For Research Use Only. Not for use in diagnostic procedures.