missing translation for 'onlineSavingsMsg'
Learn More

NAD+, MedChemExpress

Product Code. p-200100432
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16422179

Brand: Medchem Express HYB0445

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.

NAD+ is a coenzyme composed of ribosylnicotinamide 5'-diphosphate coupled to adenosine 5'-phosphate by pyrophosphate linkage.

Specifications

Chemical Name or Material NAD+
CAS 53-84-9
Color White
Purity Grade Notes Research
Molecular Formula C21H27N7O14P2
Quantity 1 g
Synonym β-DPN β-NAD β-Nicotinamide Adenine Dinucleotide
Solubility Information H2O : 41.67 mg/mL (62.81 mM; Need ultrasonic)
SMILES O[C@H]([C@@H]1O)[C@@H](O[C@@H]1COP(OP(OC[C@H]2O[C@@H]([N+]3=CC=CC(C(N)=O)=C3)[C@H](O)[C@@H]2O)([O-])=O)(O)=O)N4C(N=CN=C5N)=C5N=C4
Molecular Weight (g/mol) 663.43
Formula Weight 663.43
Percent Purity 98.24%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.