missing translation for 'onlineSavingsMsg'
Learn More

NADP disodium salt, MedChemExpress

Product Code. p-200096041
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16462326

Brand: Medchem Express HYF0002A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NADP disodium salt (Disodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP disodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

NADP disodium salt (Disodium NADP), a nicotinamide adenine dinucleotide, is a redox cofactor. NADP disodium salt is a key cofactor for electron transfer in the metabolism, being alternately oxidized (NADP+) and reduced (NADPH).

Specifications

Chemical Name or Material NADP disodium salt
CAS 24292-60-2
Color White
Purity Grade Notes Research
Molecular Formula C21H28N7O17P3.2Na
Quantity 500 mg
Synonym Disodium NADP
Solubility Information H2O : 250 mg/mL (316.70 mM; Need ultrasonic) ∣DMSO : 3.33 mg/mL (4.22 mM; ultrasonic and warming and heat to 80°C)
SMILES NC1=C2C(N([C@H]3[C@H](OP(O)(O)=O)[C@H](O)[C@@H](COP(OP([O-])(OC[C@@H]4[C@@H](O)[C@@H](O)[C@H]([N+]5=CC=CC(C(N)=O)=C5)O4)=O)(O)=O)O3)C=N2)=NC=N1.[2 Na]
Molecular Weight (g/mol) 789.38
Formula Weight 789.39
Percent Purity 98.47%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt