Learn More
BRAND™ BLAUBRAND™ Nageotte Counting Chamber
The area of 100mm² is subdivided into 40 rectangles with an area of 2.5mm² each. This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
Brand: BRAND™ 721305
This item is not returnable.
View return policy
Description
- The area of 100mm² is subdivided into 40 rectangles with an area of 2.5mm² each.
- This chamber is commonly used for counting cells in the cerebro-spinal fluid, or for counting nematodes.
- Double-ruled
- Without clips
- CE-marked according to IVD-Directive 98/79 CE.
Specifications
Counting Device | |
1Each | |
40 x 2.5mmW rectangles |
Nageotte | |
Counting cells in the cerebro-spinal fluid, or nematodes. |