missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Syringe Filters
GREENER_CHOICE

Product Code. p-4524900
Click to view available options
Pore Size:
0.2 μm
0.45 μm
0.8 μm
Diameter (Metric):
4 mm
13 mm
25 mm
Material (Membrane):
Cellulose Acetate
Glass Fiber, Cellulose Acetate
Nylon
PES
SFCA
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
Pack of 125
Pack of 50

Product Code. 15342378

Brand: Thermo Scientific™ 7201320

Only null left
Add to basket
Edge
Add to basket

Filter sample volumes of 10 to 100mL with these sterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.

Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.

Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit

Surfactant-free Cellulose Acetate (SFCA)

  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings

Cellulose Acetate (CA)

  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings

Nylon

  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certified sterile, non-cytotoxic and non-pyrogenic
Filter Type Membrane Filter
Material (Membrane) PES
Pore Size 0.2 μm
Sterility Sterile
For Use With (Application) Sterile filtration and clarification of cell culture media, buffers and additives
Housing Material Polypropylene
Quantity 100/Pk.
Type Syringe Filter
Volume (Metric) 2 to 10 mL
Diameter (Metric) 13 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.