Learn More
Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier
Secure bottles and reduce chance of bumping, cracking or chipping with this safety bottle carrier.
Brand: Thermo Scientific™ 6505-0010
150.66 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 55
Additional Details : Weight : 1.69200kg
Description
Secure bottles and reduce chance of bumping, cracking, or chipping. Thermo Scientific™ Nalgene™ 500mL LLDPE Safety Bottle Carrier offers greater rigidity and impact resistance. The sturdy, wide handle is molded in one piece with the base allowing a comfortable grip, even when the carrier is full. This linear low density polyethylene carrier holds up to six standard 500mL reagent bottles in any profile.
- Cover latches securely to base and allow carriers to stack
- Dimensions: 14L × 10W × 11″H (35 × 25 × 28cm)
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Bottle Carrier | |
Yellow | |
LLDPE | |
Yes | |
1 |
Carrier Safety Bottle LLDPE 500mL | |
Bottles (500 mL) | |
6 | |
6 |