missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Acrylic Parafilm™ Dispenser
GREENER_CHOICE

Securely hold rolls for easy dispensing with the Thermo Scientific™ Nalgene™ Acrylic Parafilm™ Dispenser.

Brand:  Thermo Scientific™ 5833-0001

71.80 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10547292

  • £112.32 / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Securely hold rolls for easy dispensing with the Thermo Scientific™ Nalgene™ Acrylic Parafilm™ Dispenser. This acrylic dispenser can hold one box of 4 in. film or two boxes of 2 in. film. A channeled front allows for cutting Parafilm.

Ordering Information:

Storage for blade is included but blade is not included.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 x 4.75 x 6 in.
121 mm
Acrylic Parafilm Dispenser
152 mm
152 mm
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Acrylic Parafilm™ Dispenser > OD, 152 x 121 x 152mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.