Learn More
Thermo Scientific™ Nalgene™ HDPE, Amber Carboy
Description
Protect photosensitive chemicals, reagents, buffers and standards when mixing and storing. Thermo Scientific™ Nalgene™ Amber HDPE Carboys with amber polypropylene (PP) closure meet the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials.
Features:
- Opaque
- Convenient wide shoulder handles
- Leakproof†
- Not Autoclavable
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Specifications
Autoclavable | Not Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Certifications/Compliance | Meets the U.S. Pharmacopoeia Light Transmission Test (USP latest edition) requirements for storage of light sensitive materials. |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Amber |
Description | Amber HDPE Carboy |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage, Transportation |
Includes | Wide shoulder handles |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1388068 | Certificate of Compliance | 08/07/2023 | 2256-7020 |
1360847 | Certificate of Compliance | 13/12/2022 | 2256-7020 |
1360016 | Certificate of Compliance | 25/05/2022 | 2256-7020 |
1333964 | Certificate of Compliance | 23/11/2021 | 2256-7020 |
1323054 | Certificate of Compliance | 13/07/2021 | 2256-7020 |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.