Learn More
Thermo Scientific™ Nalgene™ Amber PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Nonsterile
Identify samples quickly with these amber PPCO high-profile closures with color-coded inserts. Eliminate risky O-rings while providing a barrier for light-sensitive reagents.
£258.15 - £364.00
Specifications
Description | High-profile closures w/color coders for micro packaging vials |
---|---|
Diameter (Metric) Outer | 11 mm |
Material | PPCO |
Sterility | Non-sterile |
No. per Case | 1000 |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
10727283
|
Thermo Scientific™
362825-0111 |
White |
£258.15
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11304525
|
Thermo Scientific™
362825-0112 |
Yellow |
£258.15
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11314525
|
Thermo Scientific™
362825-0113 |
Orange |
£258.15
Pack of 1000 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10098772
|
Thermo Scientific™
362825-0114 |
Green |
£258.15
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
10425513
|
Thermo Scientific™
362825-0115 |
Red |
£258.15
Pack of 1000 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10592035
|
Thermo Scientific™
362825-0116 |
Blue |
£258.15
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
10374623
|
Thermo Scientific™
362825-0117 |
Gold |
£258.15
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
10028922
|
Thermo Scientific™
362825-0118 |
Purple |
£258.15
Pack of 1000 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10344103
|
Thermo Scientific™
362825-0119 |
Natural |
£258.15
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11324525
|
Thermo Scientific™
362825-1110 |
Teal |
£258.15
Pack of 1000 |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10304153
|
Thermo Scientific™
362825-1111 |
Amber |
£364.00
Pack of 1000 |
In Stock
Log in to see stock. |
|||||
Description
Identify samples quickly using Thermo Scientific™ Nalgene™ Amber PPCO High-Profile Closures with Color Coders for Micro Packaging Vials. Noncytotoxic and nonpyrogenic, these closures do not use O-rings that might fall out or contaminate contents.
- Vials (listed separately) and closures are pressure-tested together at 7.5psig.
- Screw closure is threaded.
- Permanently attached, color-coded inserts are free of toxic heavy metals.
- Inserts do not come in contact with vial contents.
- Create a leakproof† seal when used with Nalgene Micro Packaging Vials
Ordering Information:
Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1000 per case. Order nonsterile amber vials separately (Cat. No. 362805).
Specifications
High-profile closures w/color coders for micro packaging vials | |
PPCO | |
1000 | |
Nalgene | |
Screw Cap | |
ISO 13485:2016 | |
11 mm |
11 mm | |
Non-sterile | |
500 | |
PPCO | |
500 Bag, 1000 Cs. | |
Closure |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.