Learn More
Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Copolymer (PPCO) Lowboys with Spigot
Dispense most liquids, including reagents, distilled water and titrants, using these polypropylene copolymer (PPCO) lowboys, which fit into tight spaces and can stack three high.
£135.53 - £156.39
Specifications
Closure Size | 63 mm |
---|---|
Type | Carboy with Spigot |
Material | PPCO |
Diameter (English) Inner Neck | 1.73 in. |
Diameter (Metric) Inner Neck | 44 mm |
Alert:
Disengage threads and dismount spigot before autoclaving
Follow autoclave instructions provided with the product for proper function
Note: Tubing adapter is not autoclavable
Product Code | Brand | Closure Size | Capacity (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Size | Capacity (Metric) | Price | Quantity & Availability | ||||
10799091
|
Thermo Scientific™
2324-0008 |
63 mm | 8 L |
£135.53
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
||||
10662561
|
Thermo Scientific™
2324-0015 |
63 mm | 15 L |
£156.39
Each |
In Stock
Log in to see stock. |
||||
Description
Create a compact, space saving, reagent dispensing bank using chemical-resistant Thermo Scientific™ Nalgene™ Low-Profile autoclavable polypropylene copolymer (PPCO) Lowboys. The low-profile design of these high density polyethylene lowboys fits into tight spaces on shelves and in refrigerators and can stack three high. Chemically compatible with a wide range of lab liquids. Convenient integral carry handle for safe, easy transport. Nalgene polypropylene copolymer carboys can be autoclaved before refilling to eliminate bacterial growth when storing sterile solutions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables.
- Polypropylene spigot and screw closure
- Contour handle allows easy one-handed carrying — even when full
- Can tolerate strong acids and bases for a wide variety of solutions
- Follow autoclave instructions provided with the product to ensure proper function
- Leakproof† when used with Nalgene Closures;
- Carboy and Carton are recyclable in many communities
- Manufactured from non-toxic materials
- Replacement spigot sold separately Nalgene Cat. No. 6422-0010
Includes:
Polypropylene screw closures.
.
Specifications
63 mm | |
PPCO | |
44 mm | |
1 |
Carboy with Spigot | |
1.73 in. | |
4 |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.