missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Autoclavable Septum Closure, Autoclavable, 38-430 size
Click to view available options
Description:
Autoclavable Septum Closures
Description
Enable aseptic injection of reagent or sample withdrawal without compromising sterility or integrity of contents with Thermo Scientific™ Nalgene™ Autoclavable Septum Closure, suitable for use with any bottle or container with 38-430 neck, including Nalgene culture vessels.
- Use with 18-gauge or smaller needles
- Polypropylene closure with thermoplastic elastomer septum
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Description | Autoclavable Septum Closures |
Material | Polypropylene |
No. per Case | 12 |
Sterility | Non-sterile |
Closure Included | Yes |
Autoclavable | Autoclavable |
Mouth | Threaded |
Product Type | Septum Closure |
For Use With (Equipment) | Nalgene bottles with 38-430 threaded mouth. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'