missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nalgene™ Biohazardous Waste Containers

missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
5.5 L
19 L
57 L
Diameter (Metric) Outer:
22 cm
28 cm
34 cm
Height (Metric):
28 cm
43 cm
74 cm
missing translation for 'unitSize'
Each
Description
Conduct secondary containment of biohazardous waste with Thermo Scientific™ Nalgene™ Biohazardous Waste Containers, which feature convenient one-step sterilization—no need to remove the bag. A port opening in the cover allows for one-handed disposal of materials, limiting exposure to infectious waste. The containers hold standard sized autoclave bags (not included).
Features:
- Will not rust, dent or puncture like other materials
- Complies with U.S. OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens
- Autoclavable/Biohazard
- Symbol printed in English and Spanish
.
Specifications
Specifications
| Capacity (English) | 1.5 gal. |
| Certifications/Compliance | U.S. OSHA Standard 29 CFR Part 1910.1030 |
| Diameter (English) Outer | 8.75 in. |
| Capacity (Metric) | 5.5 L |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Waste Container Biohazardous |
| Height (English) | 11 in. |
| Height (Metric) | 28 cm |
| No. per Case | 1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction